accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HClH2NNH

CAS RN 1670-14-0

IUPAC Name: hydrogen benzenecarboximidamide chloride
Synonyms: hydrogen benzamidine chloride
Molecular Weight (g/mol): 156.61
Molecular Formula: C7H9ClN2
InChi Key: LZCZIHQBSCVGRD-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC(=N)C1=CC=CC=C1
Quantity 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results