accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16026-77-0

OHH2NCl

CAS RN 16026-77-0

IUPAC Name: 3-amino-4-chlorophenol
Synonyms: 3-amino-4-chlorophenol
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: FWUALUHYKLDYAN-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=CC(O)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results