accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 15996-78-8

H3NClFFF

CAS RN 15996-78-8

IUPAC Name: [2-chloro-5-(trifluoromethyl)phenyl]methanaminium
Synonyms: [2-chloro-5-(trifluoromethyl)phenyl]methanaminium
Molecular Weight (g/mol): 210.6
Molecular Formula: C8H8ClF3N
InChi Key: XOCMZYUQWVACMA-UHFFFAOYSA-O
SMILES: [NH3+]CC1=CC(=CC=C1Cl)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results