accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOOF

CAS RN 1578-70-7

IUPAC Name: ethyl 1-fluoro-2-oxocyclohexane-1-carboxylate
Synonyms: ethyl 1-fluoro-2-oxocyclohexane-1-carboxylate
Molecular Weight (g/mol): 188.2
Molecular Formula: C9H13FO3
InChi Key: WBRNDLRGLOAFCQ-UHFFFAOYNA-N
SMILES: CCOC(=O)C1(F)CCCCC1=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results