accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 156-59-2

ClCl(Z)

CAS RN 156-59-2

IUPAC Name: (1Z)-1,2-dichloroethene
Synonyms: cis-1,2-dichloroethene 1,2-dichloroethene (cis) C2H2cl2 acetalyne dichloride
Molecular Weight (g/mol): 96.94
Molecular Formula: C2H2Cl2
InChi Key: KFUSEUYYWQURPO-UPHRSURJSA-N
SMILES: Cl\C=C/Cl
Molecular Weight (g/mol) 
  • (2)
  • (2)
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results