accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 15469-97-3

IUPAC Name: 1-(triphenylmethyl)-1H-imidazole
Synonyms: 1-(triphenylmethyl)imidazole
Molecular Weight (g/mol): 310.4
Molecular Formula: C22H18N2
InChi Key: NPZDCTUDQYGYQD-UHFFFAOYSA-N
SMILES: C1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results