accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 152803-24-2

IUPAC Name: 5-hydroxypyridine-3-carbonitrile
Synonyms: 5-hydroxypyridine-3-carbonitrile
Molecular Weight (g/mol): 120.11
Molecular Formula: C6H4N2O
InChi Key: KAXIYYPIORYZLB-UHFFFAOYSA-N
SMILES: OC1=CN=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results