accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NH2NHNNN

CAS RN 152460-10-1

IUPAC Name: 6-methyl-N1-[4-(pyridin-3-yl)pyrimidin-2-yl]benzene-1,3-diamine
Synonyms: 6-methyl-N1-[4-(pyridin-3-yl)pyrimidin-2-yl]benzene-1,3-diamine
Molecular Weight (g/mol): 277.33
Molecular Formula: C16H15N5
InChi Key: QGAIPGVQJVGBIA-UHFFFAOYSA-N
SMILES: CC1=C(NC2=NC=CC(=N2)C2=CN=CC=C2)C=C(N)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results