accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

(R)HNOHOOOH3C

CAS RN 15027-14-2

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}hexanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 265.31
Molecular Formula: C14H19NO4
InChi Key: NMYWMOZOCYAHNC-UHFFFAOYNA-N
SMILES: CCCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results