accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3ONHNH2O

CAS RN 150-69-6

IUPAC Name: (4-ethoxyphenyl)urea
Synonyms: dulcin
Molecular Weight (g/mol): 180.21
Molecular Formula: C9H12N2O2
InChi Key: GGLIEWRLXDLBBF-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(NC(N)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results