accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 149022-16-2

HClH3CHNOHN

CAS RN 149022-16-2

IUPAC Name: hydrogen 1-methyl-2H,7H,9H-pyrido[3,4-b]indol-7-one chloride
Synonyms: hydrogen harmol chloride
Molecular Weight (g/mol): 234.68
Molecular Formula: C12H11ClN2O
InChi Key: RBOUBJPHXSVUTH-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=C2NC3=CC(=O)C=CC3=C2C=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results