Learn More
Harmol Hydrochloride Dihydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Harmol hydrochloride dihydrate |
| CAS | 149022-16-2 |
| Molecular Formula | C12 H10 N2 O . H Cl . 2 H2 O |
| Quantity | 500 mg |
| Synonym | 1-Methyl-9H-pyrido(3,4-b)indol-7-ol monohydrochloride dihydrate |
| InChI Formula | InChI=1S/C12H10N2O.ClH.2H2O/c1-7-12-10(4-5-13-7)9-3-2-8(15)6-11(9)14-12;;;/h2-6,14-15H,1H3;1H;2*1H2 |
| SMILES | O.O.Cl.Cc1nccc2c3ccc(O)cc3[nH]c12 |
| IUPAC Name | 1-methyl-9H-pyrido[3,4-b]indol-7-ol;dihydrate;hydrochloride |
| Molecular Weight (g/mol) | 270.71 |
| Formula Weight | 270.0771 |