accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 148-18-5

NaH3CNH3CSS

CAS RN 148-18-5

IUPAC Name: sodium (diethylcarbamothioyl)sulfanide
Synonyms: sodium diethyldithiocarbamate
Molecular Weight (g/mol): 171.25
Molecular Formula: C5H10NNaS2
InChi Key: IOEJYZSZYUROLN-UHFFFAOYSA-M
SMILES: [Na+].CCN(CC)C([S-])=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results