Learn More
Ditiocarb sodium, MedChemExpress
Product Code. 16482349
Encompass_Preferred
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. 16482349
Brand: Medchem Express HYB1637
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Ditiocarb sodium (Sodium diethyldithiocarbamate) is an accelerator of the rate of copper cementation. Sodium diethyldithiocarbamate reduces the incidence of HIV infection.
Ditiocarb sodium (Sodium diethyldithiocarbamate) is an accelerator of the rate of copper cementation. Sodium diethyldithiocarbamate reduces the incidence of HIV infection.
Specifications
Chemical Name or Material Ditiocarb sodium
Physical Form Solid
CAS 148-18-5
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C5H10NNaS2
For Use With (Application) COVID-19-anti-virus
Synonym Sodium diethyldithiocarbamate
Solubility Information DMSO : 100 mg/mL (583.91 mM; Need ultrasonic)
SMILES S=C(S[Na])N(CC)CC
Molecular Weight (g/mol) 171.26
Formula Weight 171.26
Percent Purity 98.13%
Grade Research