accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1462-37-9

BrO

CAS RN 1462-37-9

IUPAC Name: [(2-bromoethoxy)methyl]benzene
Synonyms: [(2-bromoethoxy)methyl]benzene
Molecular Weight (g/mol): 215.09
Molecular Formula: C9H11BrO
InChi Key: FWOHDAGPWDEWIB-UHFFFAOYSA-N
SMILES: BrCCOCC1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (4)