accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CSnCH3H3CH3C

CAS RN 1461-25-2

IUPAC Name: tetrabutylstannane
Synonyms: tetrabutyltin
Molecular Weight (g/mol): 347.17
Molecular Formula: C16H36Sn
InChi Key: AFCAKJKUYFLYFK-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)