CAS RN 146-80-5
CAS RN 146-80-5
IUPAC Name:
9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2,3,6,9-tetrahydro-1H-purine-2,6-dione
Synonyms:
xanthosine
Molecular Weight (g/mol):
284.23
Molecular Formula:
C10H12N4O6
InChi Key:
UBORTCNDUKBEOP-UHFFFAOYNA-N
SMILES:
OCC1OC(C(O)C1O)N1C=NC2=C1NC(=O)NC2=O
1
–
1
of
1
results
1
MedChemExpress Xanthosine
MedChemExpress Xanthosine is a nucleoside derived from xanthine and ribose. Xanthosine can increase mammary stem cell population and milk production in cattle and goats.