Learn More
MedChemExpress Xanthosine
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Molecular Weight (g/mol) | 284.23 g/mol |
| Product Type | Natural Products |
| Content And Storage | Powder -20°C 3 years∣In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Purity | 95.0% |
| Quantity | 50 mg |
| Solubility Information | DMSO : 33.33 mg/mL (117.26 mM∣Need ultrasonic) |
| For Use With (Application) | Metabolism-protein/nucleotide metabolism |
| CAS | 146-80-5 |