CAS RN 1454585-06-8
CAS RN 1454585-06-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
SR-3029, MedChemExpress
MedChemExpress SR-3029 is a potent and ATP competitive CK1δ and CK1ε inhibitor, with IC50s of 44 nM and 260 nM, respectively, and Kis of 97 nM for both kinases.