missing translation for 'onlineSavingsMsg'
Learn More

SR-3029, MedChemExpress

Product Code. p-200100546
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16499985

Brand: Medchem Express HY100011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SR-3029 is a potent and ATP competitive CK1δ and CK1ε inhibitor, with IC50s of 44 nM and 260 nM, respectively, and Kis of 97 nM for both kinases.

SR-3029 is a potent and ATP competitive CK1δ and CK1ε inhibitor, with IC50s of 44 nM and 260 nM, respectively, and Kis of 97 nM for both kinases.

Specifications

Chemical Name or Material SR-3029
CAS 1454585-06-8
Color White
Purity Grade Notes Research
Molecular Formula C23H19F3N8O
Quantity 2 mg
Solubility Information DMSO : ≥ 30 mg/mL (62.44 mM)
SMILES FC1=CC(N2C=NC3=C(NCC4=NC5=CC=C(F)C(F)=C5N4)N=C(N6CCOCC6)N=C23)=CC=C1
Molecular Weight (g/mol) 480.45
Formula Weight 480.45
Percent Purity 97.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.