accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1449144-79-9

BSOONCH3HOOH

CAS RN 1449144-79-9

IUPAC Name: {3-[benzyl(ethyl)sulfamoyl]phenyl}boronic acid
Synonyms: 3-[benzyl(ethyl)sulfamoyl]phenylboronic acid
Molecular Weight (g/mol): 319.18
Molecular Formula: C15H18BNO4S
InChi Key: QZDYSNQDEYMZBN-UHFFFAOYSA-N
SMILES: CCN(CC1=CC=CC=C1)S(=O)(=O)C1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results