CAS RN 1446089-82-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Desmethyl-N-benzyl Sildenafil, TRC
CAS: 1446089-82-2 Molecular Formula: C28 H34 N6 O4 S Molecular Weight (g/mol): 550.67 Synonym: 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[2-ethoxy-5-[[4-(phenylmethyl)-1-piperazinyl]sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-,5-[2-Ethoxy-5-[[4-(phenylmethyl)-1-piperazinyl]sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one,Benzyl Sildenafil,Sildenafil-N-benzyl IUPAC Name: 5-[5-(4-benzylpiperazin-1-yl)sulfonyl-2-ethoxyphenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one SMILES: CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(Cc5ccccc5)CC4