accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 142421-57-6

FFFPFFFFFFClFFF

CAS RN 142421-57-6

IUPAC Name: bis[3,5-bis(trifluoromethyl)phenyl](chloro)phosphane
Synonyms: bis[3,5-bis(trifluoromethyl)phenyl](chloro)phosphane
Molecular Weight (g/mol): 492.63
Molecular Formula: C16H6ClF12P
InChi Key: DFZQEHBNAJGDCT-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(=C1)P(Cl)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results