accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1421263-95-7

H3CNOFF

CAS RN 1421263-95-7

IUPAC Name: N-benzyl-N-ethyl-2,6-difluorobenzamide
Synonyms: N-benzyl-N-ethyl-2,6-difluorobenzamide
Molecular Weight (g/mol): 275.3
Molecular Formula: C16H15F2NO
InChi Key: WWMNXFJMLWOKHV-UHFFFAOYSA-N
SMILES: CCN(CC1=CC=CC=C1)C(=O)C1=C(F)C=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results