CAS RN 140-82-9
IUPAC Name:
2-[2-(diethylamino)ethoxy]ethan-1-ol
Synonyms:
2-[2-(diethylamino)ethoxy]ethanol
Molecular Weight (g/mol):
161.25
Molecular Formula:
C8H19NO2
InChi Key:
VKBVRNHODPFVHK-UHFFFAOYSA-N
SMILES:
CCN(CC)CCOCCO
1
–
2
of
2
results
1
2-(2-Diethylaminoethoxy)ethanol, TRC
CAS: 140-82-9 Molecular Formula: C8 H19 N O2 Molecular Weight (g/mol): 161.24 Synonym: 2-[2-(Diethylamino)ethoxy]ethanol,Oxeladin Hydrogen Citrate Imp. A (EP) IUPAC Name: 2-[2-(diethylamino)ethoxy]ethanol SMILES: CCN(CC)CCOCCO
2
2-[2-(Diethylamino)ethoxy]ethanol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.