accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COFOHF

CAS RN 1373920-65-0

IUPAC Name: 4-butoxy-2,6-difluorophenol
Synonyms: 4-butoxy-2,6-difluorophenol
Molecular Weight (g/mol): 202.2
Molecular Formula: C10H12F2O2
InChi Key: NZXOEPFBEOGOTJ-UHFFFAOYSA-N
SMILES: CCCCOC1=CC(F)=C(O)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results