CAS RN 1367567-95-0
CAS RN 1367567-95-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(1R)-2-(Benzylmethylamino)-1-(3-hydroxyphenyl)ethanol (Benzylphenylephrine), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
(AlphaR)-3-Hydroxy-Alpha-[[methyl(phenylmethyl)amino]methyl]-benzenemethanol, TRC
CAS: 1367567-95-0 Molecular Formula: C16 H19 N O2 Molecular Weight (g/mol): 257.33 Synonym: (1R)-2-(Benzylmethylamino)-1-(3-hydroxyphenyl)ethanol,Benzylphenylephrine,(R)-3-{2-[Benzyl(methyl)amino]-1-hydroxyethyl}phenol IUPAC Name: 3-[(1R)-2-[benzyl(methyl)amino]-1-hydroxyethyl]phenol SMILES: CN(C[C@H](O)c1cccc(O)c1)Cc2ccccc2