accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BOOCH3CH3H3CCH3N

CAS RN 1357387-29-1

IUPAC Name: 9-benzyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Synonyms: 9-benzyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Molecular Weight (g/mol): 383.3
Molecular Formula: C25H26BNO2
InChi Key: UMWJZMUSYJMTOI-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)N(CC1=CC=CC=C1)C1=C2C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results