accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OFFFFOOH(E/Z)

CAS RN 1353001-72-5

IUPAC Name: 3-[3-fluoro-5-(trifluoromethoxy)phenyl]prop-2-enoic acid
Synonyms: 3-[3-fluoro-5-(trifluoromethoxy)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 250.15
Molecular Formula: C10H6F4O3
InChi Key: KCORPQZTOLVNPS-UHFFFAOYSA-N
SMILES: OC(=O)C=CC1=CC(OC(F)(F)F)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results