accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COFOFFF

CAS RN 1352999-53-1

IUPAC Name: 1-[3-fluoro-5-(trifluoromethoxy)phenyl]ethan-1-one
Synonyms: 1-[3-fluoro-5-(trifluoromethoxy)phenyl]ethanone
Molecular Weight (g/mol): 222.14
Molecular Formula: C9H6F4O2
InChi Key: VOBVUHOUGRWDOZ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(OC(F)(F)F)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results