accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13432-81-0

ClHOClSOOCl

CAS RN 13432-81-0

IUPAC Name: 3,5-dichloro-4-hydroxybenzene-1-sulfonyl chloride
Synonyms: 3,5-dichloro-4-hydroxybenzenesulfonyl chloride
Molecular Weight (g/mol): 261.5
Molecular Formula: C6H3Cl3O3S
InChi Key: AELFLPJWVRGXKU-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=C(C=C1Cl)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results