CAS RN 1334719-95-7
CAS RN 1334719-95-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Q203, MedChemExpress
MedChemExpress Q203 (IAP6) is a midazopyridine amide compound. Q203 is active against Mycobacterium tuberculosis H37Rv with an MIC50 of 2.7 nM in culture broth medium.