Learn More
Q203, MedChemExpress
Product Code. 16499786
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16499786
Brand: Medchem Express HY101040
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Q203 (IAP6) is a midazopyridine amide compound. Q203 is active against Mycobacterium tuberculosis H37Rv with an MIC50 of 2.7 nM in culture broth medium.
Q203 (IAP6) is a midazopyridine amide compound. Q203 is active against Mycobacterium tuberculosis H37Rv with an MIC50 of 2.7 nM in culture broth medium.
Specifications
Chemical Name or Material Q203
CAS 1334719-95-7
Color White
Purity Grade Notes Research
Molecular Formula C29H28ClF3N4O2
Quantity 50 mg
Synonym IAP6 Telacebec
Solubility Information DMSO : 20 mg/mL (35.91 mM; Need ultrasonic)
SMILES O=C(C1=C(CC)N=C2C=CC(Cl)=CN21)NCC3=CC=C(N4CCC(C5=CC=C(OC(F)(F)F)C=C5)CC4)C=C3
Molecular Weight (g/mol) 557.01
Formula Weight 557.01
Percent Purity 99.45%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation