accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13304-62-6

H2CONH

CAS RN 13304-62-6

IUPAC Name: N-benzylprop-2-enamide
Synonyms: N-benzylprop-2-enamide
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: OHLHOLGYGRKZMU-UHFFFAOYSA-N
SMILES: C=CC(=O)NCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results