accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OFOCl

CAS RN 1323966-27-3

IUPAC Name: 4-chloro-3-ethoxy-2-fluorobenzaldehyde
Synonyms: 4-chloro-3-ethoxy-2-fluorobenzaldehyde
Molecular Weight (g/mol): 202.61
Molecular Formula: C9H8ClFO2
InChi Key: UHWKSKYUNZUQFC-UHFFFAOYSA-N
SMILES: CCOC1=C(Cl)C=CC(C=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results