accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3IH2N

CAS RN 13194-68-8

IUPAC Name: 4-iodo-2-methylaniline
Synonyms: 4-iodo-2-methylaniline
Molecular Weight (g/mol): 233.05
Molecular Formula: C7H8IN
InChi Key: BGKLFAQCHHCZRZ-UHFFFAOYSA-N
SMILES: CC1=CC(I)=CC=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results