accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BNOHHOHCl

CAS RN 1310403-86-1

IUPAC Name: (quinolin-6-yl)boronic acid hydrochloride
Synonyms: quinolin-6-ylboronic acid hydrochloride
Molecular Weight (g/mol): 209.44
Molecular Formula: C9H9BClNO2
InChi Key: XEUWKJJLVYUQIS-UHFFFAOYSA-N
SMILES: Cl.OB(O)C1=CC=C2N=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results