accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BrHON

CAS RN 13073-28-4

IUPAC Name: 3-bromo-2-hydroxybenzonitrile
Synonyms: 3-bromo-2-hydroxybenzonitrile
Molecular Weight (g/mol): 198.02
Molecular Formula: C7H4BrNO
InChi Key: BUWXNBPLRLEXOZ-UHFFFAOYSA-N
SMILES: OC1=C(C=CC=C1Br)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results