accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 130191-91-2

FBrFOH

CAS RN 130191-91-2

IUPAC Name: 4-bromo-3,5-difluorophenol
Synonyms: 4-bromo-3,5-difluorophenol
Molecular Weight (g/mol): 208.99
Molecular Formula: C6H3BrF2O
InChi Key: HHGOLZGZHXELSW-UHFFFAOYSA-N
SMILES: OC1=CC(F)=C(Br)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results