accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COON

CAS RN 129721-53-5

IUPAC Name: N,N-dicyclohexyl-4-methoxybenzamide
Synonyms: N,N-dicyclohexyl-4-methoxybenzamide
Molecular Weight (g/mol): 315.46
Molecular Formula: C20H29NO2
InChi Key: UBNMVLOOOQOJSF-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)N(C1CCCCC1)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results