accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1261478-61-8

CH3OFFFOH

CAS RN 1261478-61-8

IUPAC Name: [2-methyl-5-(trifluoromethoxy)phenyl]methanol
Synonyms: [2-methyl-5-(trifluoromethoxy)phenyl]methanol
Molecular Weight (g/mol): 206.16
Molecular Formula: C9H9F3O2
InChi Key: NIZSWVZPENQEME-UHFFFAOYSA-N
SMILES: CC1=C(CO)C=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results