CAS RN 1256360-37-8
IUPAC Name:
2-[3-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms:
2-[3-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol):
314.99
Molecular Formula:
C13H17BBrFO2
InChi Key:
CWASIKJVXVGLKA-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=C(F)C(CBr)=CC=C1
1
–
1
of
1
results
1
3-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 1256360-37-8 Molecular Formula: C13H17BBrFO2 Molecular Weight (g/mol): 314.989 MDL Number: MFCD17214291 InChI Key: CWASIKJVXVGLKA-UHFFFAOYSA-N Synonym: 2-3-bromomethyl-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,3-bromomethyl-2-fluorophenylboronic acid pinacol ester,3-bromomethyl-2-fluorophenylboronic acid, pinacol ester PubChem CID: 53217261 IUPAC Name: 2-[3-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane SMILES: B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)CBr)F