CAS RN 1243583-85-8
CAS RN 1243583-85-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MG 149, MedChemExpress
MedChemExpress MG149 (Tip60 HAT inhibitor) is a selective and potent Tip60 inhibitor with IC50 of 74 uM, similar potentcy for MOF (IC50= 47 uM); little potent for PCAF and p300 (IC50 >200 uM).