Learn More
MG 149, MedChemExpress
Product Code. 16462957
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16462957
Brand: Medchem Express HY15887
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress MG149 (Tip60 HAT inhibitor) is a selective and potent Tip60 inhibitor with IC50 of 74 uM, similar potentcy for MOF (IC50= 47 uM); little potent for PCAF and p300 (IC50 >200 uM).
MG149 (Tip60 HAT inhibitor) is a selective and potent Tip60 inhibitor with IC50 of 74 uM, similar potentcy for MOF (IC50= 47 uM)
Specifications
Chemical Name or Material MG 149
CAS 1243583-85-8
Color White
Purity Grade Notes Research
Molecular Formula C22H28O3
Quantity 50 mg
Synonym Tip60 HAT inhibitor
Solubility Information DMSO : ≥ 100 mg/mL (293.72 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O)C1=C(O)C=CC=C1CCC2=CC=C(CCCCCCC)C=C2
Molecular Weight (g/mol) 340.46
Formula Weight 340.46
Percent Purity 99.86%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death