accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1240257-72-0

FSFFFFFOH

CAS RN 1240257-72-0

IUPAC Name: 3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)phenol
Synonyms: 3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)phenol
Molecular Weight (g/mol): 238.15
Molecular Formula: C6H4F6OS
InChi Key: DCXIIYIPHWVVQA-UHFFFAOYSA-N
SMILES: OC1=CC(F)=CC(=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results