CAS RN 1210344-83-4
CAS RN 1210344-83-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ertugliflozin L-pyroglutamic acid, MedChemExpress
MedChemExpress Ertugliflozin L-pyroglutamic acid (PF-04971729 L-pyroglutamic acid) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.