missing translation for 'onlineSavingsMsg'
Learn More

Ertugliflozin L-pyroglutamic acid, MedChemExpress

Product Code. p-200097858
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16439127

Brand: Medchem Express HY15461A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ertugliflozin L-pyroglutamic acid (PF-04971729 L-pyroglutamic acid) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.

Ertugliflozin L-pyroglutamic acid (PF-04971729 L-pyroglutamic acid) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.

Specifications

Chemical Name or Material Ertugliflozin L-pyroglutamic acid
CAS 1210344-83-4
Color White
Purity Grade Notes Research
Molecular Formula C27H32ClNO10
Quantity 5 mg
Synonym PF-04971729 L-pyroglutamic acid
Solubility Information DMSO : ≥ 125 mg/mL (220.85 mM)
SMILES ClC1=CC=C([C@]23O[C@@](CO)(CO3)[C@@H](O)[C@H](O)[C@H]2O)C=C1CC4=CC=C(OCC)C=C4.O=C5CC[C@@H](C(O)=O)N5
Molecular Weight (g/mol) 566.0
Formula Weight 566.0
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.