CAS RN 1206593-34-1
IUPAC Name:
3-[4-ethoxy-3-(trifluoromethyl)phenyl]propanoic acid
Synonyms:
3-[4-ethoxy-3-(trifluoromethyl)phenyl]propanoic acid
Molecular Weight (g/mol):
262.23
Molecular Formula:
C12H13F3O3
InChi Key:
WDXHCCSBBUVONT-UHFFFAOYSA-N
SMILES:
CCOC1=C(C=C(CCC(O)=O)C=C1)C(F)(F)F
1
–
1
of
1
results
1
3-[4-Ethoxy-3-(trifluoromethyl)phenyl]propionic acid, 97%, Thermo Scientific™
CAS: 1206593-34-1 Molecular Formula: C12H13F3O3 Molecular Weight (g/mol): 262.228 MDL Number: MFCD13196589 InChI Key: WDXHCCSBBUVONT-UHFFFAOYSA-N Synonym: 3-4-ethoxy-3-trifluoromethyl phenyl propionic acid,3-4-ethoxy-3-trifluoromethyl phenyl propanoic acid PubChem CID: 57361410 IUPAC Name: 3-[4-ethoxy-3-(trifluoromethyl)phenyl]propanoic acid SMILES: CCOC1=C(C=C(C=C1)CCC(=O)O)C(F)(F)F