accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 12019-52-2

CuTeCu

CAS RN 12019-52-2

IUPAC Name: (cupriotellanyl)copper
Synonyms: (cupriotellanyl)copper
Molecular Weight (g/mol): 254.69
Molecular Formula: Cu2Te
InChi Key: MZEWONGNQNXVKA-UHFFFAOYSA-N
SMILES: [Cu][Te][Cu]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results