accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1198-27-2

ClNH3HO

CAS RN 1198-27-2

IUPAC Name: 2-hydroxynaphthalen-1-aminium chloride
Synonyms: 2-hydroxynaphthalen-1-aminium chloride
Molecular Weight (g/mol): 195.65
Molecular Formula: C10H10ClNO
InChi Key: DEKREBCFNLUULC-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]C1=C2C=CC=CC2=CC=C1O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)